siRNA Details
| Gene Symbol: MFSD4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153369.2 | NP_699200.2 | 4 | 1864 | ||
| XM_017011483.1 | XP_016866972.1 | 3 | 1747 | |||
| GenBank mRNA | AB067506.1 | - | 1 | 1099 | ||
| AF161363.1 | - | 1 | 178 | |||
| AK093136.1 | - | 4 | 1823 | |||
| AK292735.1 | - | 4 | 1793 | |||
| AK315016.1 | - | 4 | 1788 | |||
| BC036115.1 | - | 4 | 1773 | 5298999 | ||

MFSD4B
Genomic Map