siRNA Details
| Gene Symbol: INTS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033547.3 | NP_291025.3 | 5 | 663 | ||
| XM_011545352.2 | XP_011543654.1 | 6 | 832 | |||
| XM_011545353.2 | XP_011543655.1 | 5 | 841 | |||
| XM_017018560.1 | XP_016874049.1 | 5 | 696 | |||
| GenBank mRNA | AK290866.1 | - | 5 | 659 | ||
| AL834171.1 | - | 5 | 606 | |||
| BC008013.1 | - | 1 | 116 | |||
| BC009995.2 | - | 5 | 658 | 4129693 | ||
| BC015664.2 | - | 4 | 556 | 4554197 | ||

INTS4
Genomic Map