siRNA Details
| Gene Symbol: INTS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033547.3 | NP_291025.3 | 6, 7 | 728 | ||
| XM_011545352.2 | XP_011543654.1 | 7, 8 | 897 | |||
| XM_011545353.2 | XP_011543655.1 | 6, 7 | 906 | |||
| XM_017018560.1 | XP_016874049.1 | 6, 7 | 761 | |||
| GenBank mRNA | AK290866.1 | - | 6, 7 | 724 | ||
| AL834171.1 | - | 6, 7 | 671 | |||
| BC008013.1 | - | 2, 3 | 181 | |||
| BC009995.2 | - | 6, 7 | 723 | 4129693 | ||
| BC015664.2 | - | 5, 6 | 621 | 4554197 | ||

INTS4
Genomic Map