siRNA Details
| Gene Symbol: MTG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138384.2 | NP_612393.2 | 4 | 405 | ||
| GenBank mRNA | AK074976.1 | - | 4 | 326 | ||
| AK125012.1 | - | 5 | 692 | |||
| AK292515.1 | - | 4 | 247 | |||
| AK293988.1 | - | 4 | 313 | |||
| AK308997.1 | - | 4 | 354 | |||
| BC000920.2 | - | 2 | 114 | |||
| BC004409.1 | - | 4 | 364 | |||
| BC026039.1 | - | 4 | 381 | 5444745 | ||
| BC026725.1 | - | 4 | 284 | |||
| BC035721.1 | - | 4 | 309 | 5588933 | ||
| DB136242.1 | - | 4 | 405 | |||

MTG1
Genomic Map