siRNA Details
| Gene Symbol: MTG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138384.2 | NP_612393.2 | 6 | 537 | ||
| GenBank mRNA | AK074976.1 | - | 6 | 458 | ||
| AK125012.1 | - | 7 | 824 | |||
| AK292515.1 | - | 6 | 379 | |||
| AK308997.1 | - | 6 | 557 | |||
| BC000920.2 | - | 4 | 246 | |||
| BC004409.1 | - | 6 | 496 | |||
| BC026039.1 | - | 6 | 513 | 5444745 | ||
| BC026725.1 | - | 6 | 416 | |||
| BC035721.1 | - | 6 | 441 | 5588933 | ||

MTG1
Genomic Map