siRNA Details
| Gene Symbol: UBTD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152277.2 | NP_689490.2 | 3 | 772 | ||
| XM_017010022.1 | XP_016865511.1 | 3 | 782 | |||
| XM_017010023.1 | XP_016865512.1 | 3 | 706 | |||
| GenBank mRNA | AF251700.1 | - | 1 | 294 | ||
| AK022894.1 | - | 1 | 302 | |||
| AK313274.1 | - | 3 | 738 | |||
| BC019910.1 | - | 3 | 620 | 4402070 | ||
| BU849695.1 | - | 3 | 787 | |||

UBTD2
Genomic Map