siRNA Details
| Gene Symbol: RCSD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001322923.1 | NP_001309852.1 | 4 | 468 | ||
| NM_001322924.1 | NP_001309853.1 | 3 | 396 | |||
| NM_052862.3 | NP_443094.3 | 5 | 514 | |||
| GenBank mRNA | AF545852.1 | - | 5 | 658 | ||
| AL137762.1 | - | 5 | 435 | |||
| AL833211.1 | - | 3 | 204 | |||
| AY530954.1 | - | 4 | 327 | |||
| BC101536.1 | - | 5 | 351 | 8069042 | ||
| BC101562.1 | - | 5 | 351 | 8069068 | ||
| BC143428.1 | - | 4 | 261 | 9051936 | ||
| DR006615.1 | - | 5 | 541 | |||

RCSD1
Genomic Map