siRNA Details
| Gene Symbol: CHMP4C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152284.3 | NP_689497.1 | 2 | 477 | ||
| GenBank mRNA | AB120734.1 | - | 2 | 309 | ||
| AK000049.1 | - | 2 | 439 | |||
| AK314873.1 | - | 2 | 476 | |||
| AY329086.1 | - | 2 | 311 | |||
| BC014321.1 | - | 2 | 481 | 3828410 | ||
| BX492886.1 | - | 2 | 497 | |||

CHMP4C
Genomic Map