siRNA Details
| Gene Symbol: RBM33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053043.2 | NP_444271.2 | 13 | 2639 | ||
| XM_005249531.1 | XP_005249588.1 | 14 | 2804 | |||
| XM_005249532.1 | XP_005249589.1 | 14 | 2804 | |||
| XM_005249533.1 | XP_005249590.1 | 14 | 2774 | |||
| XM_005249534.1 | XP_005249591.1 | 13 | 2684 | |||
| XM_005249535.1 | XP_005249592.1 | 13 | 2654 | |||
| XM_011515840.1 | XP_011514142.1 | 14 | 2804 | |||
| GenBank mRNA | AK301179.1 | - | 8 | 1586 | ||
| AK307515.1 | - | 13 | 2469 | |||
| AL137724.1 | - | 11 | 3534 | |||

RBM33
Genomic Map