siRNA Details
| Gene Symbol: MGME1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001310338.1 | NP_001297267.1 | 4 | 902 | ||
| NM_052865.3 | NP_443097.1 | 3 | 857 | |||
| XM_005260867.2 | XP_005260924.1 | 4 | 1124 | |||
| XM_005260870.3 | XP_005260927.1 | 3 | 805 | |||
| XM_006723663.3 | XP_006723726.1 | 4 | 850 | |||
| XM_011529394.1 | XP_011527696.1 | 4 | 739 | |||
| XM_011529395.1 | XP_011527697.1 | 3 | 617 | |||
| XM_017028127.1 | XP_016883616.1 | 3 | 1079 | |||
| XM_017028128.1 | XP_016883617.1 | 3 | 953 | |||
| GenBank mRNA | AF306684.1 | - | 3 | 666 | ||
| AF306685.1 | - | 3 | 666 | |||
| AK027503.1 | - | 3 | 661 | |||
| AK310282.1 | - | 2 | 173 | |||
| AK315532.1 | - | 3 | 693 | |||
| BC016869.1 | - | 3 | 629 | 3850853 | ||
| BX382492.2 | - | 3 | 667 | |||

MGME1
Genomic Map