siRNA Details
| Gene Symbol: HELB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033647.4 | NP_387467.2 | 4 | 870 | ||
| NR_135080.1 | 4 | 870 | ||||
| NR_135081.1 | 4 | 870 | ||||
| XM_005269234.2 | XP_005269291.1 | 4 | 848 | |||
| XM_017020230.1 | XP_016875719.1 | 4 | 848 | |||
| GenBank mRNA | AF319995.1 | - | 4 | 811 | ||
| AK290894.1 | - | 4 | 835 | |||
| BC143409.1 | - | 4 | 836 | |||
| BC143411.1 | - | 4 | 836 | |||

HELB
Genomic Map