siRNA Details
| Gene Symbol: SHKBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138392.3 | NP_612401.2 | 4 | 281 | ||
| XM_006723474.1 | XP_006723537.1 | 4 | 281 | |||
| XM_006723475.1 | XP_006723538.1 | 4 | 281 | |||
| XM_011527492.1 | XP_011525794.1 | 4 | 281 | |||
| XM_011527493.1 | XP_011525795.1 | 3 | 146 | |||
| XM_017027473.1 | XP_016882962.1 | 4 | 281 | |||
| XM_017027474.1 | XP_016882963.1 | 3 | 146 | |||
| GenBank mRNA | AF258553.1 | - | 4 | 280 | ||
| AK075057.1 | - | 4 | 277 | |||
| AK123695.1 | - | 2 | 259 | |||
| AK300808.1 | - | 3 | 219 | |||
| AK300819.1 | - | 4 | 277 | |||
| AK312748.1 | - | 4 | 245 | |||
| BC022855.1 | - | 4 | 252 | 4102901 | ||
| DB038927.1 | - | 4 | 281 | |||

SHKBP1
Genomic Map