siRNA Details
Gene Symbol: CCDC102A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033212.3 | NP_149989.2 | 4 | 1116 | ||
XM_011523469.2 | XP_011521771.1 | 4 | 2396 | |||
GenBank mRNA | BC004307.1 | - | 1 | 73 | ||
BC008285.1 | - | 4 | 1095 | 3637387 | ||
BC009941.2 | - | 4 | 1116 | 4100726 |
