siRNA Details
| Gene Symbol: MARCH9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138396.5 | NP_612405.2 | 4 | 1209 | ||
| GenBank mRNA | AK093897.1 | - | 2 | 124 | ||
| AK313925.1 | - | 2 | 886 | |||
| BC036455.2 | - | 4 | 1190 | 4811410 | ||
| BC050397.1 | - | 4 | 1209 | |||

MARCH9
Genomic Map