siRNA Details
Gene Symbol: COQ10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099337.1 | NP_001092807.1 | 4 | 522 | ||
NM_144576.3 | NP_653177.3 | 4 | 766 | |||
GenBank mRNA | AK057003.1 | - | 4 | 682 | ||
AK057014.1 | - | 3 | 483 | |||
AY358861.1 | - | 4 | 454 | |||
BC002435.2 | - | 3 | 343 | |||
BC026922.1 | - | 4 | 522 | |||
BC047444.1 | - | 4 | 494 | |||
BC073923.1 | - | 4 | 650 | 5188138 |
