siRNA Details
| Gene Symbol: MVB12A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304547.1 | NP_001291476.1 | 7 | 812 | ||
| NM_138401.3 | NP_612410.1 | 6 | 885 | |||
| GenBank mRNA | AK057057.1 | - | 7 | 1370 | ||
| AK289487.1 | - | 6 | 685 | |||
| BC007883.2 | - | 6 | 621 | 3688605 | ||
| BC011840.2 | - | 6 | 629 | 4053084 | ||
| BG684013.1 | - | 7 | 777 | |||
| BY996463.1 | - | 4 | 446 | |||

MVB12A
Genomic Map