siRNA Details
| Gene Symbol: ARMC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199196.1 | NP_001186125.1 | 4, 5 | 702 | ||
| NM_033415.3 | NP_219483.1 | 3, 4 | 594 | |||
| XM_005260157.3 | XP_005260214.1 | 3, 4 | 645 | |||
| XM_017027484.1 | XP_016882973.1 | 3, 4 | 569 | |||
| XM_017027485.1 | XP_016882974.1 | 5, 6 | 826 | |||
| GenBank mRNA | AK295488.1 | - | 3, 4 | 368 | ||
| BC056161.1 | - | 4, 5 | 551 | |||
| BC067125.1 | - | 4, 5 | 471 | 6161112 | ||
| BX648486.1 | - | 3, 4 | 489 | |||
| Z36779.1 | - | 2, 3 | 109 | |||

ARMC6
Genomic Map