siRNA Details
| Gene Symbol: TPTE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001141968.1 | NP_001135440.1 | 11, 12 | 954 | ||
| NM_130785.3 | NP_570141.3 | 10, 11 | 864 | |||
| NM_199254.2 | NP_954863.2 | 13, 14 | 1095 | |||
| NR_073485.1 | 15, 16 | 1207 | ||||
| NR_073486.1 | 8, 9 | 534 | ||||
| GenBank mRNA | AJ421032.1 | - | 10, 11 | 864 | ||
| AJ421033.1 | - | 7, 8 | 491 | |||
| AK301741.1 | - | 15, 16 | 1207 | |||
| AY219890.1 | - | 13, 14 | 1095 | |||
| BC128146.1 | - | 11, 12 | 936 | 40112547 | ||

TPTE2
Genomic Map