siRNA Details
| Gene Symbol: HTRA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297559.1 | NP_001284488.1 | 6 | 1157 | ||
| NM_053044.4 | NP_444272.1 | 6 | 1157 | |||
| XM_011513596.2 | XP_011511898.1 | 6 | 1165 | |||
| GenBank mRNA | AY040094.2 | - | 6 | 1174 | ||
| AY280665.1 | - | 6 | 1145 | |||
| AY280666.1 | - | 6 | 1060 | |||
| BC034390.1 | - | 6 | 1157 | 5180585 | ||
| BC035717.1 | - | 6 | 1157 | 5588608 | ||

HTRA3
Genomic Map