siRNA Details
Gene Symbol: CEP41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018718.2 | NP_061188.1 | 9, 10 | 992 | ||
NR_046443.1 | 9, 10 | 916 | ||||
XM_011516709.2 | XP_011515011.1 | 10, 11 | 953 | |||
XM_011516710.2 | XP_011515012.1 | 8, 9 | 768 | |||
GenBank mRNA | AF429308.1 | - | 9, 10 | 838 | ||
AJ278890.1 | - | 9, 10 | 795 | |||
AK026098.1 | - | 9, 10 | 792 | |||
AK311382.1 | - | 9, 10 | 733 | |||
AK314676.1 | - | 9, 10 | 771 | |||
AL359617.1 | - | 3, 4 | 293 | |||
AY186739.1 | - | 1, 2 | 434 |
