siRNA Details
| Gene Symbol: MRAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138409.2 | NP_612418.2 | 4 | 667 | ||
| XM_011535400.2 | XP_011533702.1 | 4 | 593 | |||
| XM_017010219.1 | XP_016865708.1 | 5 | 965 | |||
| XM_017010220.1 | XP_016865709.1 | 4 | 1234 | |||
| GenBank mRNA | AK090775.1 | - | 4 | 667 | ||
| AK292736.1 | - | 4 | 598 | |||
| BC010003.2 | - | 4 | 612 | 4134852 | ||
| BC039855.1 | - | 4 | 530 | 3850270 | ||

MRAP2
Genomic Map