siRNA Details
| Gene Symbol: STX1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052874.4 | NP_443106.1 | 3 | 374 | ||
| XM_017022893.1 | XP_016878382.1 | 3 | 129 | |||
| GenBank mRNA | AY028792.1 | - | 3 | 190 | ||
| AY995211.1 | - | 3 | 132 | |||
| BC062298.1 | - | 3 | 186 | 6375788 | ||
| BX102270.1 | - | 4 | 268 | |||
| DN990202.1 | - | 3 | 368 | |||
| HY332651.1 | - | 3 | 411 | |||

STX1B
Genomic Map