siRNA Details
| Gene Symbol: GLMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256604.1 | NP_001243533.1 | 4 | 808 | ||
| NM_001256605.1 | NP_001243534.1 | 3 | 550 | |||
| NM_001256608.1 | NP_001243537.1 | 4 | 808 | |||
| NM_001256609.1 | NP_001243538.1 | 4 | 1037 | |||
| NM_144580.2 | NP_653181.1 | 4 | 808 | |||
| XM_011509117.2 | XP_011507419.1 | 3 | 467 | |||
| XM_017000149.1 | XP_016855638.1 | 4 | 804 | |||
| GenBank mRNA | AK075349.1 | - | 4 | 777 | ||
| AK296157.1 | - | 4 | 1005 | |||
| AL528533.3 | - | 3 | 522 | |||
| AY358450.1 | - | 4 | 781 | |||
| BC011575.1 | - | 4 | 791 | |||
| BC018757.1 | - | 4 | 777 | 4850051 | ||
| BC036340.1 | - | 4 | 724 | |||
| BM833126.1 | - | 2 | 210 | |||
| BP302818.1 | - | 2 | 275 | |||
| CN479212.1 | - | 2 | 570 | |||
| CX756681.1 | - | 4 | 723 | |||

GLMP
Genomic Map