siRNA Details
| Gene Symbol: SGF29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138414.2 | NP_612423.1 | 2 | 210 | ||
| XM_017022894.1 | XP_016878383.1 | 2 | 223 | |||
| GenBank mRNA | AA132818.1 | - | 2 | 211 | ||
| AK057008.1 | - | 2 | 169 | |||
| BC011981.1 | - | 2 | 201 | 3863665 | ||

SGF29
Genomic Map