siRNA Details
| Gene Symbol: SGF29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138414.2 | NP_612423.1 | 10 | 963 | ||
| GenBank mRNA | AA132314.1 | - | 1 | 193 | ||
| AK057008.1 | - | 10 | 922 | |||
| BC011981.1 | - | 10 | 954 | 3863665 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SGF29| Gene Symbol: SGF29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138414.2 | NP_612423.1 | 10 | 963 | ||
| GenBank mRNA | AA132314.1 | - | 1 | 193 | ||
| AK057008.1 | - | 10 | 922 | |||
| BC011981.1 | - | 10 | 954 | 3863665 | ||