siRNA Details
| Gene Symbol: PLCD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133373.4 | NP_588614.1 | 6 | 1185 | ||
| XM_011524253.2 | XP_011522555.1 | 6 | 1170 | |||
| GenBank mRNA | AB075844.1 | - | 6 | 967 | ||
| AK074240.1 | - | 6 | 1166 | |||
| AL834392.1 | - | 6 | 1027 | |||
| BC010668.1 | - | 4 | 547 | |||
| BC072384.1 | - | 6 | 1150 | 6378601 | ||

PLCD3
Genomic Map