siRNA Details
Gene Symbol: GLCCI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138426.3 | NP_612435.1 | 8 | 1985 | ||
GenBank mRNA | AK055741.1 | - | 1 | 1386 | ||
AK126731.1 | - | 2 | 1136 | |||
AL832860.1 | - | 2 | 97 | |||
AL833694.1 | - | 4 | 480 | |||
BC011254.1 | - | 2 | 193 | |||
BC050291.1 | - | 8 | 1936 | 5272003 |
