siRNA Details
| Gene Symbol: HELQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297755.1 | NP_001284684.1 | 7 | 1700 | ||
| NM_001297756.1 | NP_001284685.1 | 8 | 1914 | |||
| NM_001297757.1 | NP_001284686.1 | 7 | 1803 | |||
| NM_133636.3 | NP_598375.2 | 8 | 1901 | |||
| XM_005262711.1 | XP_005262768.1 | 8 | 1762 | |||
| XM_005262713.2 | XP_005262770.1 | 8 | 1991 | |||
| XM_006714076.2 | XP_006714139.1 | 8 | 1863 | |||
| XM_011531580.2 | XP_011529882.1 | 8 | 1863 | |||
| XM_017007679.1 | XP_016863168.1 | 7 | 1561 | |||
| XM_017007680.1 | XP_016863169.1 | 7 | 1790 | |||
| XM_017007681.1 | XP_016863170.1 | 8 | 1863 | |||
| XM_017007682.1 | XP_016863171.1 | 8 | 1982 | |||
| XM_017007683.1 | XP_016863172.1 | 6 | 1101 | |||
| GenBank mRNA | AF436845.1 | - | 8 | 1862 | ||
| AK308395.1 | - | 7 | 1640 | |||
| AK312725.1 | - | 8 | 1878 | |||
| AL512702.1 | - | 6 | 565 | |||
| BC111502.1 | - | 7 | 1723 | |||
| CK000270.1 | - | 5 | 501 | |||

HELQ
Genomic Map