siRNA Details
| Gene Symbol: SLC35A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080670.3 | NP_542401.1 | 3 | 1177 | ||
| GenBank mRNA | AF445025.1 | - | 2 | 2430 | ||
| AJ420598.1 | - | 1 | 190 | |||
| AK289378.1 | - | 1 | 531 | |||
| AL833972.1 | - | 2 | 896 | |||
| BC009906.2 | - | 3 | 907 | 2961449 | ||

SLC35A4
Genomic Map