siRNA Details
| Gene Symbol: SLC22A9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080866.2 | NP_543142.2 | 3 | 850 | ||
| XM_017017159.1 | XP_016872648.1 | 3 | 1768 | |||
| GenBank mRNA | AB062418.1 | - | 3 | 683 | ||
| AB074812.1 | - | 3 | 831 | |||
| AF440402.1 | - | 3 | 818 | |||
| AJ295270.1 | - | 3 | 642 | |||
| AK074246.1 | - | 3 | 850 | |||
| BC126288.1 | - | 3 | 732 | 8992004 | ||

SLC22A9
Genomic Map