siRNA Details
| Gene Symbol: PGLYRP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052891.2 | NP_443123.1 | 7 | 1036 | ||
| XM_011509118.1 | XP_011507420.1 | 9 | 1465 | |||
| XM_011509119.1 | XP_011507421.1 | 8 | 1357 | |||
| XM_011509120.2 | XP_011507422.1 | 9 | 1243 | |||
| GenBank mRNA | AY035376.1 | - | 7 | 1036 | ||
| BC069741.1 | - | 7 | 1022 | 7262478 | ||
| BC069798.1 | - | 7 | 1023 | 7262490 | ||
| BC069801.1 | - | 7 | 1023 | 7262502 | ||
| BC128114.1 | - | 7 | 1009 | 40112227 | ||
| BC128115.1 | - | 7 | 1009 | 40112231 | ||

PGLYRP3
Genomic Map