siRNA Details
| Gene Symbol: PGLYRP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052891.2 | NP_443123.1 | 6 | 862 | ||
| XM_011509118.1 | XP_011507420.1 | 8 | 1291 | |||
| XM_011509119.1 | XP_011507421.1 | 7 | 1183 | |||
| XM_011509120.2 | XP_011507422.1 | 8 | 1069 | |||
| GenBank mRNA | AY035376.1 | - | 6 | 862 | ||
| BC069741.1 | - | 6 | 848 | 7262478 | ||
| BC069798.1 | - | 6 | 849 | 7262490 | ||
| BC069801.1 | - | 6 | 849 | 7262502 | ||
| BC128114.1 | - | 6 | 835 | 40112227 | ||
| BC128115.1 | - | 6 | 835 | 40112231 | ||

PGLYRP3
Genomic Map