siRNA Details
| Gene Symbol: PNMA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001103150.1 | NP_001096620.1 | 2 | 1091 | ||
| NM_001103151.1 | NP_001096621.1 | 3 | 991 | |||
| NM_001184924.1 | NP_001171853.1 | 4 | 1076 | |||
| NM_052926.2 | NP_443158.1 | 2 | 1077 | |||
| XM_017029252.1 | XP_016884741.1 | 4 | 1393 | |||
| XM_017029253.1 | XP_016884742.1 | 3 | 1055 | |||
| GenBank mRNA | AB067521.1 | - | 2 | 1074 | ||
| AK295446.1 | - | 3 | 955 | |||
| AK316104.1 | - | 3 | 960 | |||
| AY121806.1 | - | 2 | 1074 | |||
| BC101111.1 | - | 2 | 935 | 40011824 | ||
| BC101112.2 | - | 2 | 921 | 40011826 | ||
| BC101113.1 | - | 2 | 921 | 40011827 | ||

PNMA5
Genomic Map