siRNA Details
| Gene Symbol: OSBPL1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242508.1 | NP_001229437.1 | 9 | 949 | ||
| NM_018030.4 | NP_060500.3 | 7 | 1318 | |||
| NM_080597.3 | NP_542164.2 | 21 | 2205 | |||
| XM_006722380.3 | XP_006722443.1 | 20 | 2118 | |||
| XM_006722382.3 | XP_006722445.1 | 20 | 2083 | |||
| XM_017025530.1 | XP_016881019.1 | 21 | 2497 | |||
| XM_017025533.1 | XP_016881022.1 | 7 | 801 | |||
| XM_017025534.1 | XP_016881023.1 | 7 | 3329 | |||
| GenBank mRNA | AF274714.1 | - | 7 | 1318 | ||
| AF323726.2 | - | 21 | 2093 | |||
| AF392449.1 | - | 7 | 939 | |||
| AF392450.1 | - | 21 | 2162 | |||
| AK001079.1 | - | 8 | 846 | |||
| AK096358.1 | - | 21 | 2205 | |||
| AK292895.1 | - | 22 | 2137 | |||
| AK301862.1 | - | 9 | 949 | |||
| AK314552.1 | - | 7 | 769 | |||
| BC063420.1 | - | 7 | 1004 | |||
| BC073854.1 | - | 5 | 525 | |||
| BC098384.1 | - | 7 | 997 | |||
| BC136452.1 | - | 21 | 2102 | 9020441 | ||
| BC144117.1 | - | 21 | 2102 | 9052637 | ||

OSBPL1A
Genomic Map