siRNA Details
| Gene Symbol: OSBPL1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242508.1 | NP_001229437.1 | 13 | 1394 | ||
| NM_018030.4 | NP_060500.3 | 11 | 1763 | |||
| NM_080597.3 | NP_542164.2 | 25 | 2650 | |||
| XM_006722380.3 | XP_006722443.1 | 24 | 2563 | |||
| XM_006722382.3 | XP_006722445.1 | 24 | 2528 | |||
| XM_017025530.1 | XP_016881019.1 | 25 | 2942 | |||
| XM_017025531.1 | XP_016881020.1 | 23 | 2252 | |||
| XM_017025533.1 | XP_016881022.1 | 11 | 1246 | |||
| XM_017025534.1 | XP_016881023.1 | 11 | 3774 | |||
| GenBank mRNA | AF274714.1 | - | 11 | 1763 | ||
| AF323726.2 | - | 25 | 2538 | |||
| AF392449.1 | - | 11 | 1384 | |||
| AK096358.1 | - | 25 | 2650 | |||
| AK292895.1 | - | 26 | 2582 | |||
| AK301862.1 | - | 13 | 1394 | |||
| AK314552.1 | - | 11 | 1214 | |||
| BC063420.1 | - | 11 | 1449 | |||
| BC098384.1 | - | 11 | 1442 | |||
| BC136452.1 | - | 25 | 2547 | 9020441 | ||
| BC144117.1 | - | 25 | 2547 | 9052637 | ||

OSBPL1A
Genomic Map