siRNA Details
| Gene Symbol: OSBPL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003712.1 | NP_001003712.1 | 16 | 2199 | ||
| NM_001319652.1 | NP_001306581.1 | 19 | 2324 | |||
| NM_001319653.1 | NP_001306582.1 | 17 | 2236 | |||
| NM_001319655.1 | NP_001306584.1 | 14 | 2130 | |||
| NM_020841.4 | NP_065892.1 | 17 | 2245 | |||
| XM_005268621.4 | XP_005268678.1 | 20 | 2343 | |||
| XM_006719224.2 | XP_006719287.1 | 16 | 2204 | |||
| XM_011537855.2 | XP_011536157.1 | 17 | 1862 | |||
| XM_011537856.2 | XP_011536158.1 | 18 | 2280 | |||
| XM_011537857.2 | XP_011536159.1 | 20 | 2334 | |||
| XM_017018768.1 | XP_016874257.1 | 17 | 1877 | |||
| XM_017018769.1 | XP_016874258.1 | 18 | 2248 | |||
| GenBank mRNA | AB040884.3 | - | 17 | 1916 | ||
| AF392452.1 | - | 17 | 2246 | |||
| AK092160.1 | - | 14 | 2131 | |||
| AK289997.1 | - | 17 | 1965 | |||
| AL832876.1 | - | 4 | 301 | |||
| BC012935.1 | - | 3 | 230 | |||
| BC082235.1 | - | 17 | 2184 | |||
| BC093834.1 | - | 16 | 1844 | 7939679 | ||
| BC101529.1 | - | 16 | 1844 | 8069035 | ||
| BC111728.1 | - | 17 | 2139 | 40029118 | ||
| BX647211.1 | - | 16 | 1801 | |||
| CR749542.1 | - | 19 | 2049 | |||

OSBPL8
Genomic Map