siRNA Details
| Gene Symbol: TMEM123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052932.2 | NP_443164.2 | 4 | 930 | ||
| GenBank mRNA | AK075420.1 | - | 4 | 714 | ||
| AK225790.1 | - | 4 | 901 | |||
| AY008283.1 | - | 3 | 668 | |||
| AY157580.1 | - | 4 | 625 | |||
| AY358696.1 | - | 4 | 602 | |||
| BC032296.1 | - | 4 | 617 | 4340763 | ||

TMEM123
Genomic Map