siRNA Details
| Gene Symbol: ZNF554 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102651.1 | NP_001096121.1 | 3, 4 | 460 | ||
| XM_011527660.2 | XP_011525962.1 | 3, 4 | 301 | |||
| XM_011527662.2 | XP_011525964.1 | 2, 3 | 203 | |||
| GenBank mRNA | AK092136.1 | - | 4, 5 | 512 | ||
| AK299752.1 | - | 3, 4 | 449 | |||
| BC000113.1 | - | 3, 4 | 371 | |||
| BC047777.1 | - | 3, 4 | 429 | 6187000 | ||
| DC319914.1 | - | 3, 4 | 461 | |||

ZNF554
Genomic Map