siRNA Details
| Gene Symbol: GBP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134486.2 | NP_001127958.1 | 5 | 996 | ||
| NM_052942.3 | NP_443174.1 | 6 | 1104 | |||
| GenBank mRNA | AF288815.1 | - | 4 | 567 | ||
| AF328727.1 | - | 6 | 1067 | |||
| AF430642.1 | - | 6 | 1105 | |||
| AF430643.1 | - | 5 | 985 | |||
| AK090479.1 | - | 4 | 3816 | |||
| AK315064.1 | - | 6 | 715 | |||
| AY358953.1 | - | 5 | 930 | |||
| BC031639.1 | - | 6 | 1091 | 5173241 | ||
| BC033761.1 | - | 2 | 252 | |||

GBP5
Genomic Map