siRNA Details
| Gene Symbol: UHRF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152896.2 | NP_690856.1 | 3 | 842 | ||
| NR_046386.1 | 3 | 842 | ||||
| XM_011517704.2 | XP_011516006.1 | 3 | 471 | |||
| XM_017014253.1 | XP_016869742.1 | 3 | 845 | |||
| XM_017014254.1 | XP_016869743.1 | 3 | 471 | |||
| GenBank mRNA | AB071698.1 | - | 3 | 540 | ||
| AF274049.1 | - | 3 | 670 | |||
| AK308433.1 | - | 3 | 794 | |||
| BC028397.2 | - | 3 | 842 | 4797830 | ||

UHRF2
Genomic Map