siRNA Details
| Gene Symbol: ZNF689 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138447.2 | NP_612456.1 | 3 | 987 | ||
| NR_073480.1 | 3 | 622 | ||||
| NR_073481.1 | 3 | 625 | ||||
| NR_073482.1 | 3 | 505 | ||||
| NR_104166.1 | 3 | 524 | ||||
| GenBank mRNA | AB065282.1 | - | 3 | 975 | ||
| AK074896.1 | - | 3 | 710 | |||
| AK310459.1 | - | 3 | 544 | |||
| BC014000.2 | - | 3 | 962 | 3936339 | ||
| CD243730.1 | - | 1 | 172 | |||
| CN406120.1 | - | 3 | 636 | |||

ZNF689
Genomic Map