siRNA Details
| Gene Symbol: TTC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001310135.1 | NP_001297064.1 | 31 | 6096 | ||
| XM_011537430.2 | XP_011535732.1 | 27 | 3928 | |||
| XM_011537431.2 | XP_011535733.1 | 25 | 3585 | |||
| XM_017021254.1 | XP_016876743.1 | 30 | 5946 | |||
| XM_017021255.1 | XP_016876744.1 | 29 | 5811 | |||
| XM_017021256.1 | XP_016876745.1 | 30 | 6210 | |||
| GenBank mRNA | BC014342.2 | - | 4 | 503 | ||
| BC103914.2 | - | 12 | 1445 | 40006180 | ||
| BC103915.1 | - | 12 | 1445 | 40006182 | ||
| BC103916.2 | - | 12 | 1445 | 40006183 | ||
| BX161415.1 | - | 11 | 1380 | |||

TTC6
Genomic Map