siRNA Details
| Gene Symbol: TTC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001310135.1 | NP_001297064.1 | 29, 30 | 5876 | ||
| XM_011537430.2 | XP_011535732.1 | 25, 26 | 3708 | |||
| XM_011537431.2 | XP_011535733.1 | 23, 24 | 3365 | |||
| XM_017021254.1 | XP_016876743.1 | 28, 29 | 5726 | |||
| XM_017021255.1 | XP_016876744.1 | 27, 28 | 5591 | |||
| XM_017021256.1 | XP_016876745.1 | 28, 29 | 5990 | |||
| GenBank mRNA | BC014342.2 | - | 2, 3 | 283 | ||
| BC103914.2 | - | 10, 11 | 1225 | 40006180 | ||
| BC103915.1 | - | 10, 11 | 1225 | 40006182 | ||
| BC103916.2 | - | 10, 11 | 1225 | 40006183 | ||
| BX161415.1 | - | 9, 10 | 1160 | |||

TTC6
Genomic Map