siRNA Details
| Gene Symbol: DHRS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136050.2 | NP_001129522.1 | 3, 4 | 578 | ||
| NM_138452.2 | NP_612461.1 | 3, 4 | 570 | |||
| GenBank mRNA | AF418205.1 | - | 3, 4 | 514 | ||
| AK058159.1 | - | 3, 4 | 570 | |||
| AL833917.1 | - | 3, 4 | 473 | |||
| BC014057.2 | - | 3, 4 | 484 | 3622913 | ||
| BC015943.1 | - | 3, 4 | 381 | 3922816 | ||
| BX247980.1 | - | 3, 4 | 494 | |||

DHRS1
Genomic Map