siRNA Details
| Gene Symbol: CCDC151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001302453.1 | NP_001289382.1 | 13 | 1583 | ||
| NM_001302454.1 | NP_001289383.1 | 11 | 1697 | |||
| NM_145045.4 | NP_659482.3 | 13 | 1877 | |||
| GenBank mRNA | AK056363.1 | - | 13 | 1798 | ||
| AK294355.1 | - | 15 | 1777 | |||
| AK302113.1 | - | 13 | 1584 | |||
| AK308072.1 | - | 11 | 1659 | |||
| BC014252.2 | - | 12 | 1434 | |||
| BC141828.1 | - | 13 | 1809 | 40148425 | ||
| BC142637.1 | - | 13 | 1822 | 40147788 | ||
| BC144421.1 | - | 11 | 1356 | |||

CCDC151
Genomic Map