siRNA Details
| Gene Symbol: DIRC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052952.2 | NP_443184.1 | 2 | 432 | ||
| XM_017003281.1 | XP_016858770.1 | 2 | 253 | |||
| GenBank mRNA | AY039011.1 | - | 2 | 433 | ||
| BC125137.1 | - | 1 | 189 | 40119557 | ||

DIRC1
Genomic Map