siRNA Details
| Gene Symbol: ZNF526 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001314033.1 | NP_001300962.1 | 3 | 629 | ||
| NM_133444.2 | NP_597701.1 | 3 | 381 | |||
| GenBank mRNA | AB075831.1 | - | 3 | 312 | ||
| AK122649.1 | - | 3 | 305 | |||
| BC013013.2 | - | 3 | 303 | 3531734 | ||
| DA236474.1 | - | 3 | 527 | |||
| DA762298.1 | - | 3 | 381 | |||
| JQ313906.1 | - | 1 | 156 | |||

ZNF526
Genomic Map