siRNA Details
| Gene Symbol: ZNF526 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001314033.1 | NP_001300962.1 | 3 | 1133 | ||
| NM_133444.2 | NP_597701.1 | 3 | 885 | |||
| GenBank mRNA | AB075831.1 | - | 3 | 816 | ||
| AK122649.1 | - | 3 | 809 | |||
| AK298895.1 | - | 4 | 429 | |||
| AL833399.1 | - | 1 | 35 | |||
| BC013013.2 | - | 3 | 807 | 3531734 | ||
| JQ313906.1 | - | 1 | 660 | |||

ZNF526
Genomic Map