siRNA Details
| Gene Symbol: FMO9P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002925.2 | 4 | 472 | |||
| GenBank mRNA | BC014341.1 | - | 4 | 476 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FMO9P| Gene Symbol: FMO9P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002925.2 | 4 | 472 | |||
| GenBank mRNA | BC014341.1 | - | 4 | 476 | ||