siRNA Details
| Gene Symbol: WDR92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256476.1 | NP_001243405.1 | 1, 2 | 292 | ||
| NM_138458.3 | NP_612467.1 | 1, 2 | 292 | |||
| GenBank mRNA | AK056303.1 | - | 1, 2 | 256 | ||
| AY541587.1 | - | 1, 2 | 267 | |||
| BC014022.2 | - | 1, 2 | 214 | 4309224 | ||
| BC020271.1 | - | 1, 2 | 246 | |||
| BC066657.1 | - | 1, 2 | 242 | 6164689 | ||
| BP271204.1 | - | 1, 2 | 292 | |||

WDR92
Genomic Map